RIE156324945 Rieke Metals, Inc.

2,3-dichloro-3'-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]benzophenone

  • MDL number MFCD03842215
  • Cas Number : 898762-19-1
  • Molecular Weight : 406.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0600-53

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0600-53
Availability details for
Quantity
  • Availability
  • Add to Cart