RIE156324954 Rieke Metals, Inc.

cyclopropyl 3-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]phenyl ketone

  • MDL number MFCD03842224
  • Cas Number : 898762-46-4
  • Molecular Weight : 301.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0601-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0601-00
Availability details for
Quantity
  • Availability
  • Add to Cart