RIE156324958 Rieke Metals, Inc.

ethyl 4-[3-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]phenyl]-4-oxobutyrate

  • MDL number MFCD03842227
  • Cas Number : 898762-58-8
  • Molecular Weight : 361.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0605-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0605-00
Availability details for
Quantity
  • Availability
  • Add to Cart