RIE156324959 Rieke Metals, Inc.

ethyl 5-[3-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]phenyl]-5-oxovalerate

  • MDL number MFCD03842228
  • Cas Number : 898762-61-3
  • Molecular Weight : 375.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0606-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0606-00
Availability details for
Quantity
  • Availability
  • Add to Cart