RIE156324966 Rieke Metals, Inc.

4'-methyl-3-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842235
  • Cas Number : 898762-76-0
  • Molecular Weight : 311.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-methylphenyl)[3-(4-thiomorpholinylmethyl)phenyl]methanone
Inchi Code 1S/C19H21NOS/c1-15-5-7-17(8-6-15)19(21)18-4-2-3-16(13-18)14-20-9-11-22-12-10-20/h2-8,13H,9-12,14H2,1H3
Inchi Key CKHGSSYIUKJBHX-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0700-04

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0700-04
Availability details for
Quantity
  • Availability
  • Add to Cart