RIE156324969 Rieke Metals, Inc.

4'-methoxy-3-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842238
  • Cas Number : 898762-82-8
  • Molecular Weight : 327.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0700-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0700-07
Availability details for
Quantity
  • Availability
  • Add to Cart