RIE156324974 Rieke Metals, Inc.

3-carboethoxy-3'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842243
  • Cas Number : 898762-92-0
  • Molecular Weight : 369.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-[3-(4-thiomorpholinylmethyl)benzoyl]benzoate
Inchi Code 1S/C21H23NO3S/c1-2-25-21(24)19-8-4-7-18(14-19)20(23)17-6-3-5-16(13-17)15-22-9-11-26-12-10-22/h3-8,13-14H,2,9-12,15H2,1H3
Inchi Key VTGQXLUYKVHLAQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0700-12

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0700-12
Availability details for
Quantity
  • Availability
  • Add to Cart