RIE156324977 Rieke Metals, Inc.

4'-thiomethyl-3-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842246
  • Cas Number : 898762-98-6
  • Molecular Weight : 343.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0700-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0700-15
Availability details for
Quantity
  • Availability
  • Add to Cart