RIE156324982 Rieke Metals, Inc.

3-fluoro-3'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842251
  • Cas Number : 898763-07-0
  • Molecular Weight : 315.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-fluorophenyl)[3-(4-thiomorpholinylmethyl)phenyl]methanone
Inchi Code 1S/C18H18FNOS/c19-17-6-2-5-16(12-17)18(21)15-4-1-3-14(11-15)13-20-7-9-22-10-8-20/h1-6,11-12H,7-10,13H2
Inchi Key SZMJAWQJLQFVNJ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0700-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0700-22
Availability details for
Quantity
  • Availability
  • Add to Cart