RIE156324986 Rieke Metals, Inc.

2,5-dimethyl-3'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842255
  • Cas Number : 898763-16-1
  • Molecular Weight : 325.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,5-dimethylphenyl)[3-(4-thiomorpholinylmethyl)phenyl]methanone
Inchi Code 1S/C20H23NOS/c1-15-6-7-16(2)19(12-15)20(22)18-5-3-4-17(13-18)14-21-8-10-23-11-9-21/h3-7,12-13H,8-11,14H2,1-2H3
Inchi Key GOJVWAGXQZAJPY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0700-28

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0700-28
Availability details for
Quantity
  • Availability
  • Add to Cart