RIE156324987 Rieke Metals, Inc.

2,6-dimethyl-3'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842256
  • Cas Number : 898763-19-4
  • Molecular Weight : 325.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0700-29

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0700-29
Availability details for
Quantity
  • Availability
  • Add to Cart