RIE156324990 Rieke Metals, Inc.

4-bromo-3-fluoro-3'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842259
  • Cas Number : 898763-28-5
  • Molecular Weight : 394.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0700-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0700-32
Availability details for
Quantity
  • Availability
  • Add to Cart