RIE156324993 Rieke Metals, Inc.

2-chloro-3'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842262
  • Cas Number : 898763-37-6
  • Molecular Weight : 331.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-chlorophenyl)[3-(4-thiomorpholinylmethyl)phenyl]methanone
Inchi Code 1S/C18H18ClNOS/c19-17-7-2-1-6-16(17)18(21)15-5-3-4-14(12-15)13-20-8-10-22-11-9-20/h1-7,12H,8-11,13H2
Inchi Key QFFCLSAZAPATOC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0700-36

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0700-36
Availability details for
Quantity
  • Availability
  • Add to Cart