RIE156324997 Rieke Metals, Inc.

3-thiomorpholinomethyl-4'-trifluoromethylbenzophenone

  • MDL number MFCD03842266
  • Cas Number : 898763-49-0
  • Molecular Weight : 365.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0700-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0700-45
Availability details for
Quantity
  • Availability
  • Add to Cart