RIE156325003 Rieke Metals, Inc.

2,4-Dichloro-3'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842272
  • Cas Number : 898787-81-0
  • Molecular Weight : 366.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0700-54

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0700-54
Availability details for
Quantity
  • Availability
  • Add to Cart