RIE156325004 Rieke Metals, Inc.

2,5-dichloro-3'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842273
  • Cas Number : 898787-84-3
  • Molecular Weight : 366.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,5-dichlorophenyl)[3-(4-thiomorpholinylmethyl)phenyl]methanone
Inchi Code 1S/C18H17Cl2NOS/c19-15-4-5-17(20)16(11-15)18(22)14-3-1-2-13(10-14)12-21-6-8-23-9-7-21/h1-5,10-11H,6-9,12H2
Inchi Key WJNAWOOAERWDKA-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0700-55

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0700-55
Availability details for
Quantity
  • Availability
  • Add to Cart