RIE156325008 Rieke Metals, Inc.

3,4-difluoro-3'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842277
  • Cas Number : 898787-96-7
  • Molecular Weight : 333.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0700-59

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0700-59
Availability details for
Quantity
  • Availability
  • Add to Cart