RIE156325009 Rieke Metals, Inc.

3,5-difluoro-3'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842278
  • Cas Number : 898787-99-0
  • Molecular Weight : 333.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,5-difluorophenyl)[3-(4-thiomorpholinylmethyl)phenyl]methanone
Inchi Code 1S/C18H17F2NOS/c19-16-9-15(10-17(20)11-16)18(22)14-3-1-2-13(8-14)12-21-4-6-23-7-5-21/h1-3,8-11H,4-7,12H2
Inchi Key IGYFJRHAKYXQNI-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0700-60

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0700-60
Availability details for
Quantity
  • Availability
  • Add to Cart