RIE156325011 Rieke Metals, Inc.

cyclopropyl 3-(thiomorpholinomethyl)phenyl ketone

  • MDL number MFCD03842280
  • Cas Number : 898788-05-1
  • Molecular Weight : 261.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0701-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0701-00
Availability details for
Quantity
  • Availability
  • Add to Cart