RIE156325015 Rieke Metals, Inc.

ethyl 4-oxo-4-[3-(thiomorpholinomethyl)phenyl]butyrate

  • MDL number MFCD03842284
  • Cas Number : 898788-17-5
  • Molecular Weight : 321.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-oxo-4-[3-(4-thiomorpholinylmethyl)phenyl]butanoate
Inchi Code 1S/C17H23NO3S/c1-2-21-17(20)7-6-16(19)15-5-3-4-14(12-15)13-18-8-10-22-11-9-18/h3-5,12H,2,6-11,13H2,1H3
Inchi Key OXHWWDKZKZANPS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0705-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0705-00
Availability details for
Quantity
  • Availability
  • Add to Cart