RIE156325016 Rieke Metals, Inc.

ethyl 5-oxo-5-[3-(thiomorpholinomethyl)phenyl]valerate

  • MDL number MFCD03842285
  • Cas Number : 898788-20-0
  • Molecular Weight : 335.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0706-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0706-00
Availability details for
Quantity
  • Availability
  • Add to Cart