RIE156325018 Rieke Metals, Inc.

ethyl 7-oxo-7-[3-(thiomorpholinomethyl)phenyl]heptanoate

  • MDL number MFCD03842287
  • Cas Number : 898788-24-4
  • Molecular Weight : 363.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 7-oxo-7-[3-(4-thiomorpholinylmethyl)phenyl]heptanoate
Inchi Code 1S/C20H29NO3S/c1-2-24-20(23)10-5-3-4-9-19(22)18-8-6-7-17(15-18)16-21-11-13-25-14-12-21/h6-8,15H,2-5,9-14,16H2,1H3
Inchi Key SUCBSSMVKPZQHH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0708-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0708-00
Availability details for
Quantity
  • Availability
  • Add to Cart