RIE156325020 Rieke Metals, Inc.

3-(4-methylpiperazinomethyl)benzophenone

  • MDL number MFCD03842289
  • Cas Number : 898788-28-8
  • Molecular Weight : 294.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0800-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0800-01
Availability details for
Quantity
  • Availability
  • Add to Cart