RIE156325024 Rieke Metals, Inc.

2-methoxy-3'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842293
  • Cas Number : 898788-36-8
  • Molecular Weight : 324.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0800-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0800-05
Availability details for
Quantity
  • Availability
  • Add to Cart