RIE156325025 Rieke Metals, Inc.

3-methoxy-3'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842294
  • Cas Number : 898788-38-0
  • Molecular Weight : 324.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-methoxyphenyl){3-[(4-methyl-1-piperazinyl)methyl]phenyl}methanone
Inchi Code 1S/C20H24N2O2/c1-21-9-11-22(12-10-21)15-16-5-3-6-17(13-16)20(23)18-7-4-8-19(14-18)24-2/h3-8,13-14H,9-12,15H2,1-2H3
Inchi Key QRAVFDCRVNTLGI-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0800-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0800-06
Availability details for
Quantity
  • Availability
  • Add to Cart