RIE156325026 Rieke Metals, Inc.

4'-methoxy-3-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842295
  • Cas Number : 898788-40-4
  • Molecular Weight : 324.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-methoxyphenyl){3-[(4-methyl-1-piperazinyl)methyl]phenyl}methanone
Inchi Code 1S/C20H24N2O2/c1-21-10-12-22(13-11-21)15-16-4-3-5-18(14-16)20(23)17-6-8-19(24-2)9-7-17/h3-9,14H,10-13,15H2,1-2H3
Inchi Key SPYHDZVXFDWARS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0800-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0800-07
Availability details for
Quantity
  • Availability
  • Add to Cart