RIE156325027 Rieke Metals, Inc.

2-cyano-3'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842296
  • Cas Number : 898788-42-6
  • Molecular Weight : 319.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0800-08

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0800-08
Availability details for
Quantity
  • Availability
  • Add to Cart