RIE156325030 Rieke Metals, Inc.

2-carboethoxy-3'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842299
  • Cas Number : 898788-48-2
  • Molecular Weight : 366.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0800-11

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0800-11
Availability details for
Quantity
  • Availability
  • Add to Cart