RIE156325032 Rieke Metals, Inc.

4'-carboethoxy-3-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842301
  • Cas Number : 898788-52-8
  • Molecular Weight : 366.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0800-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0800-13
Availability details for
Quantity
  • Availability
  • Add to Cart