RIE156325033 Rieke Metals, Inc.

3'-(4-methylpiperazinomethyl)-2-thiomethylbenzophenone

  • MDL number MFCD03842302
  • Cas Number : 898788-54-0
  • Molecular Weight : 340.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name {3-[(4-methyl-1-piperazinyl)methyl]phenyl}[2-(methylsulfanyl)phenyl]methanone
Inchi Code 1S/C20H24N2OS/c1-21-10-12-22(13-11-21)15-16-6-5-7-17(14-16)20(23)18-8-3-4-9-19(18)24-2/h3-9,14H,10-13,15H2,1-2H3
Inchi Key IMWQWKUHCPGPOF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0800-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0800-14
Availability details for
Quantity
  • Availability
  • Add to Cart