RIE156325034 Rieke Metals, Inc.

3-(4-methylpiperazinomethyl)-4'-thiomethylbenzophenone

  • MDL number MFCD03842303
  • Cas Number : 898788-56-2
  • Molecular Weight : 340.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0800-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0800-15
Availability details for
Quantity
  • Availability
  • Add to Cart