RIE156325037 Rieke Metals, Inc.

3-Chloro-3'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842306
  • Cas Number : 898788-62-0
  • Molecular Weight : 328.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0800-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0800-20
Availability details for
Quantity
  • Availability
  • Add to Cart