RIE156325038 Rieke Metals, Inc.

4'-Chloro-3-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842307
  • Cas Number : 898788-64-2
  • Molecular Weight : 328.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-chlorophenyl){3-[(4-methyl-1-piperazinyl)methyl]phenyl}methanone
Inchi Code 1S/C19H21ClN2O/c1-21-9-11-22(12-10-21)14-15-3-2-4-17(13-15)19(23)16-5-7-18(20)8-6-16/h2-8,13H,9-12,14H2,1H3
Inchi Key FWPZHDNSTAWRHQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0800-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0800-21
Availability details for
Quantity
  • Availability
  • Add to Cart