RIE156325039 Rieke Metals, Inc.

3-fluoro-3'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842308
  • Cas Number : 898788-66-4
  • Molecular Weight : 312.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0800-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0800-22
Availability details for
Quantity
  • Availability
  • Add to Cart