RIE156325043 Rieke Metals, Inc.

2,5-dimethyl-3'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842312
  • Cas Number : 898788-74-4
  • Molecular Weight : 322.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,5-dimethylphenyl){3-[(4-methyl-1-piperazinyl)methyl]phenyl}methanone
Inchi Code 1S/C21H26N2O/c1-16-7-8-17(2)20(13-16)21(24)19-6-4-5-18(14-19)15-23-11-9-22(3)10-12-23/h4-8,13-14H,9-12,15H2,1-3H3
Inchi Key MNUBMNSIWHPROC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0800-28

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0800-28
Availability details for
Quantity
  • Availability
  • Add to Cart