RIE156325049 Rieke Metals, Inc.

3-chloro-4-fluoro-3'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842318
  • Cas Number : 898788-92-6
  • Molecular Weight : 346.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0800-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0800-34
Availability details for
Quantity
  • Availability
  • Add to Cart