RIE156325051 Rieke Metals, Inc.

2-fluoro-3'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842319
  • Cas Number : 898788-98-2
  • Molecular Weight : 312.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0800-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0800-37
Availability details for
Quantity
  • Availability
  • Add to Cart