RIE156325053 Rieke Metals, Inc.

3-(4-methylpiperazinomethyl)-3'-trifluoromethylbenzophenone

  • MDL number MFCD03842321
  • Cas Number : 898789-03-2
  • Molecular Weight : 362.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name {3-[(4-methyl-1-piperazinyl)methyl]phenyl}[3-(trifluoromethyl)phenyl]methanone
Inchi Code 1S/C20H21F3N2O/c1-24-8-10-25(11-9-24)14-15-4-2-5-16(12-15)19(26)17-6-3-7-18(13-17)20(21,22)23/h2-7,12-13H,8-11,14H2,1H3
Inchi Key ODWYKRKEGBTIQT-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0800-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0800-44
Availability details for
Quantity
  • Availability
  • Add to Cart