RIE156325061 Rieke Metals, Inc.

2,5-dichloro-3'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842329
  • Cas Number : 898789-25-8
  • Molecular Weight : 363.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0800-55

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0800-55
Availability details for
Quantity
  • Availability
  • Add to Cart