RIE156325065 Rieke Metals, Inc.

3,4-difluoro-3'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03842333
  • Cas Number : 898789-33-8
  • Molecular Weight : 330.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0800-59

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0800-59
Availability details for
Quantity
  • Availability
  • Add to Cart