RIE156325069 Rieke Metals, Inc.

cyclobutyl 3-(4-methylpiperazinomethyl)phenyl ketone

  • MDL number MFCD03842337
  • Cas Number : 898789-41-8
  • Molecular Weight : 272.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name cyclobutyl{3-[(4-methyl-1-piperazinyl)methyl]phenyl}methanone
Inchi Code 1S/C17H24N2O/c1-18-8-10-19(11-9-18)13-14-4-2-7-16(12-14)17(20)15-5-3-6-15/h2,4,7,12,15H,3,5-6,8-11,13H2,1H3
Inchi Key ZISQRXXBHWPEIE-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0802-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0802-00
Availability details for
Quantity
  • Availability
  • Add to Cart