RIE156325071 Rieke Metals, Inc.

cyclohexyl 3-(4-methylpiperazinomethyl)phenyl ketone

  • MDL number MFCD03842339
  • Cas Number : 898789-45-2
  • Molecular Weight : 300.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0804-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0804-00
Availability details for
Quantity
  • Availability
  • Add to Cart