RIE156325073 Rieke Metals, Inc.

Ethyl 5-[3-(4-methylpiperazinomethyl)phenyl]-5-oxovalerate

  • MDL number MFCD03842341
  • Cas Number : 898789-49-6
  • Molecular Weight : 332.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0806-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0806-00
Availability details for
Quantity
  • Availability
  • Add to Cart