RIE156325076 Rieke Metals, Inc.

ethyl 8-[3-(4-methylpiperazinomethyl)phenyl]-8-oxooctanoate

  • MDL number MFCD03842344
  • Cas Number : 898789-55-4
  • Molecular Weight : 374.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0809-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0809-00
Availability details for
Quantity
  • Availability
  • Add to Cart