RIE156325080 Rieke Metals, Inc.

4'-methyl-3-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842348
  • Cas Number : 898789-63-4
  • Molecular Weight : 277.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [3-(2,5-dihydro-1H-pyrrol-1-ylmethyl)phenyl](4-methylphenyl)methanone
Inchi Code 1S/C19H19NO/c1-15-7-9-17(10-8-15)19(21)18-6-4-5-16(13-18)14-20-11-2-3-12-20/h2-10,13H,11-12,14H2,1H3
Inchi Key SFYRFVRPZUYSGY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6125-0900-04

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0900-04
Availability details for
Quantity
  • Availability
  • Add to Cart