RIE156325084 Rieke Metals, Inc.

2-cyano-3'-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842352
  • Cas Number : 898789-72-5
  • Molecular Weight : 288.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0900-08

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0900-08
Availability details for
Quantity
  • Availability
  • Add to Cart