RIE156325086 Rieke Metals, Inc.

4'-cyano-3-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842354
  • Cas Number : 898789-78-1
  • Molecular Weight : 288.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0900-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0900-10
Availability details for
Quantity
  • Availability
  • Add to Cart