RIE156325087 Rieke Metals, Inc.

2-carboethoxy-3'-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842355
  • Cas Number : 898789-81-6
  • Molecular Weight : 335.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0900-11

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0900-11
Availability details for
Quantity
  • Availability
  • Add to Cart