RIE156325090 Rieke Metals, Inc.

3'-(3-pyrrolinomethyl)-2-thiomethylbenzophenone

  • MDL number MFCD03842358
  • Cas Number : 898789-90-7
  • Molecular Weight : 309.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0900-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0900-14
Availability details for
Quantity
  • Availability
  • Add to Cart