RIE156325094 Rieke Metals, Inc.

3-chloro-3'-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03842362
  • Cas Number : 898790-01-7
  • Molecular Weight : 297.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6125-0900-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6125-0900-20
Availability details for
Quantity
  • Availability
  • Add to Cart